APO455838274 Apollo Scientific Ltd

4,4,4-Trifluoro-DL-valine

  • MDL number MFCD00021721
  • Cas Number : 16063-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 245-248
Synonym 2-Amino-3-methyl-4,4,4-trifluorobutanoic acid, 2-Amino-3-methyl-4,4,4-trifluorobutyric acid
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/444-trifluoro-dl-valine-97

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/444-trifluoro-dl-valine-97
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart