APO455838281 Apollo Scientific Ltd

alpha-Methyl-3,5-bis(trifluoromethyl)benzyl alcohol

  • MDL number MFCD01632521
  • Cas Number : 368-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-74
Purity 99
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=37728

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=37728
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart