APO455838282 Apollo Scientific Ltd

4,5-Difluoro-2-methylbenzoic acid

  • MDL number MFCD08062377
  • Cas Number : 183237-86-7
  • Molecular Weight : 172.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 117-121
Iupac Name 4,5-difluoro-2-methylbenzoic acid
Inchi Code 1S/C8H6F2O2/c1-4-2-6(9)7(10)3-5(4)8(11)12/h2-3H,1H3,(H,11,12)
Inchi Key LDMHYYGJZWDBQL-UHFFFAOYSA-N

Description

Synonym 2-Carboxy-4,5-difluorotoluene, 4,5-Difluoro-o-toluic acid
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 117-121
Product Link https://store.apolloscientific.co.uk/product/45-difluoro-2-methylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/45-difluoro-2-methylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart