APO455838284 Apollo Scientific Ltd

1-Bromo-2,3-dimethyl-4-fluorobenzene

  • MDL number MFCD03789115
  • Cas Number : 52548-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=37753

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=37753
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart