APO455838295 Apollo Scientific Ltd

4-Bromo-2,5-difluorophenol

  • MDL number MFCD04973751
  • Cas Number : 486424-36-6
  • Molecular Weight : 208.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44-53
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-25-difluorophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-25-difluorophenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart