APO455838301 Apollo Scientific Ltd

3-Hydroxy-5-(trifluoromethoxy)benzeneboronic acid

  • MDL number MFCD09475898
  • Cas Number : 957120-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-81
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39402

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=39402
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart