APO455838306 Apollo Scientific Ltd

4,5-Dichloro-2-fluorobenzenesulphonyl chloride

  • MDL number MFCD03094453
  • Cas Number : 13656-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-57.5
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2974

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=2974
pictograms GHS05;GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart