APO455838308 Apollo Scientific Ltd

1-[3-(4-Fluorophenoxy)prop-1-yl]piperazine dihydrochloride

  • MDL number MFCD03428570
  • Cas Number : 537037-78-8
  • Molecular Weight : 311.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212-214
Synonym 1-Fluoro-4-[3-(piperazin-1-yl)propoxy]benzene dihydrochloride
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-3-4-fluorophenoxyprop-1-ylpiperazine-dihydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-3-4-fluorophenoxyprop-1-ylpiperazine-dihydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart