APO455838312 Apollo Scientific Ltd

2-Sulphanyl-6-(trifluoromethyl)pyridine

  • MDL number MFCD09864680
  • Cas Number : 121307-80-0
  • Molecular Weight : 179.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Mercapto-6-(trifluoromethyl)pyridine, 6-(Trifluoromethyl)pyridine-2-thiol
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-sulphanyl-6-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-sulphanyl-6-trifluoromethylpyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart