APO455838316 Apollo Scientific Ltd

3-Bromo-2-hydroxy-5-(trifluoromethyl)pyridine

  • MDL number MFCD02691223
  • Cas Number : 76041-73-1
  • Molecular Weight : 242

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-171
Synonym 3-Bromo-5-(trifluoromethyl)pyridin-2-ol, 5-Bromo-6-hydroxy-alpha,alpha,alpha-trifluoro-3-picoline
Purity 98
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-hydroxy-5-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-hydroxy-5-trifluoromethylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart