APO455838320 Apollo Scientific Ltd

2-Fluoro-3-methoxybenzyl alcohol

  • MDL number MFCD09743596
  • Cas Number : 178974-59-9
  • Molecular Weight : 156.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-59
Synonym 2-Fluoro-3-(hydroxymethyl)anisole
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 126-129/5mm
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-3-methoxybenzyl-alcohol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-3-methoxybenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart