APO455838326 Apollo Scientific Ltd

3-Chloro-5-hydroxybenzotrifluoride

  • MDL number MFCD04115852
  • Cas Number : 570391-18-3
  • Molecular Weight : 196.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Chloro-5-(trifluoromethyl)phenol, 5-Chloro-alpha,alpha,alpha-trifluoro-m-cresol
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-5-hydroxybenzotrifluoride

Safety Information

signalword Danger
pcodes P260;P280;P310
MSDS https://store.apolloscientific.co.uk/product/3-chloro-5-hydroxybenzotrifluoride
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart