APO455838327 Apollo Scientific Ltd

1-(Trifluorovinyl)cyclohex-1-ene

  • MDL number MFCD00156079
  • Cas Number : 243863-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5635

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=5635
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart