APO455838329 Apollo Scientific Ltd

2,6-Difluorobenzeneboronic acid, pinacol ester

  • MDL number MFCD09801043
  • Cas Number : 863868-37-5
  • Molecular Weight : 240.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(2,6-Difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Purity 98
Storage temp. Store at 2-8
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-difluorobenzeneboronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-difluorobenzeneboronic-acid-pinacol-ester
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart