APO455838339 Apollo Scientific Ltd

6-Bromo-1,1,2-trifluorohex-1-ene

  • MDL number MFCD01075257
  • Cas Number : 126828-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.4
Flash Point 66
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=647

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=647
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart