APO455838342 Apollo Scientific Ltd

5-Methoxy-2-(trifluoromethyl)pyridine

  • MDL number MFCD09864699
  • Cas Number : 216766-13-1
  • Molecular Weight : 177.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Methoxy-alpha,alpha,alpha-trifluoro-2-picoline
Purity 98
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-methoxy-2-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-methoxy-2-trifluoromethylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart