APO455838343 Apollo Scientific Ltd

3-Amino-2-bromobenzotrifluoride

  • MDL number MFCD01656033
  • Cas Number : 58458-10-9
  • Molecular Weight : 240.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-3-(trifluoromethyl)aniline, 2-Bromo-alpha,alpha,alpha-trifluoro-m-toluidine
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-2-bromobenzotrifluoride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-2-bromobenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart