APO455838348 Apollo Scientific Ltd

Perfluoro(1-chloro-6-iodohexane)

  • MDL number MFCD01320787
  • Cas Number : 16486-97-8
  • Molecular Weight : 462.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Chloro-1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-6-iodohexane
Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 68/48mm
Product Link https://store.apolloscientific.co.uk/product/perfluoro1-chloro-6-iodohexane

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/perfluoro1-chloro-6-iodohexane
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart