APO455838349 Apollo Scientific Ltd

2-Chloro-3-ethoxy-6-fluorobenzeneboronic acid

  • MDL number MFCD09800880
  • Cas Number : 957120-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Borono-2-chloro-4-fluorophenyl ethyl ether, 3-Borono-2-chloro-4-fluorophenetole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=41271

Safety Information

signalword Warning
pcodes P280;P301;P312;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=41271
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart