APO455838350 Apollo Scientific Ltd

2,4-Dichloro-6-(3-fluorophenoxy)-1,3,5-triazine

  • MDL number MFCD05154988
  • Cas Number : 112748-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8816

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=8816
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart