APO455838352 Apollo Scientific Ltd

3-Chloro-4-nitrobenzotrifluoride

  • MDL number MFCD00272564
  • Cas Number : 402-11-9
  • Molecular Weight : 225.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-4-(trifluoromethyl)nitrobenzene, 2-Chloro-1-nitro-4-(trifluoromethyl)benzene, 3-Chloro-4-nitro-alpha,alpha,alpha-trifluorotoluene
Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 54/ 0.4mm
Product Link https://store.apolloscientific.co.uk/product/3-chloro-4-nitrobenzotrifluoride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-4-nitrobenzotrifluoride
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart