APO455838353 Apollo Scientific Ltd

4-Fluoro-2-(trifluoromethyl)cinnamic acid

  • MDL number MFCD00236297
  • Cas Number : 243977-21-1
  • Molecular Weight : 234.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181-183
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-2-trifluoromethylcinnamic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-2-trifluoromethylcinnamic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart