APO455838354 Apollo Scientific Ltd

Ethyl 4-hydroxy-8-(trifluoromethyl)quinoline-3-carboxylate

  • MDL number MFCD00173387
  • Cas Number : 23851-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Ethoxycarbonyl)-4-hydroxy-8-(trifluoromethyl)quinoline, 3-(Ethoxycarbonyl)-4-hydroxy-8-(trifluoromethyl)-1-azanaphthalene
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/ethyl-4-hydroxy-8-trifluoromethylquinoline-3-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-4-hydroxy-8-trifluoromethylquinoline-3-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart