APO455838355 Apollo Scientific Ltd

2'-Fluoro-4'-hydroxyacetophenone

  • MDL number MFCD00142713
  • Cas Number : 98619-07-9
  • Molecular Weight : 154.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-123
Synonym 1-(2-Fluoro-4-hydroxyphenyl)ethan-1-one, 4-Acetyl-3-fluorophenol
Flash Point 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-4-hydroxyacetophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-4-hydroxyacetophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart