APO455838356 Apollo Scientific Ltd

1-(2-Chloro-6-fluorobenzyl)-5-cyano-6-methyl-1,2,3,4-tetrahydropyridin-2-one

  • MDL number MFCD00975042
  • Cas Number : 338748-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109-111
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2657

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=2657
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart