APO455838357 Apollo Scientific Ltd

2-Chloro-6-(4-fluorophenyl)nicotinonitrile

  • MDL number MFCD00215072
  • Cas Number : 31776-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173-176
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2734

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2734
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart