APO455838359 Apollo Scientific Ltd

4-Bromo-2,5-difluoronitrobenzene

  • MDL number MFCD00042184
  • Cas Number : 167415-27-2
  • Molecular Weight : 237.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-60
Synonym 1-Bromo-2,5-difluoro-4-nitrobenzene
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-25-difluoronitrobenzene

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-25-difluoronitrobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart