APO455838363 Apollo Scientific Ltd

5-Chloro-2-fluorophenetole

  • MDL number MFCD00672980
  • Cas Number : 289039-34-5
  • Molecular Weight : 174.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-2-ethoxy-1-fluorobenzene
Inchi Code 1S/C8H8ClFO/c1-2-11-8-5-6(9)3-4-7(8)10/h3-5H,2H2,1H3
Inchi Key SYVFNABINSSJKA-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-chloro-2-fluorophenetole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-2-fluorophenetole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart