APO455838364 Apollo Scientific Ltd

3-Chloro-5-fluorophenol

  • MDL number MFCD00042212
  • Cas Number : 202982-70-5
  • Molecular Weight : 146.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-5-fluorophenol

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-5-fluorophenol
pictograms GHS05;GHS07;GHS09
hstatements H302+H312;H314;H411
Availability details for
Quantity
  • Availability
  • Add to Cart