APO455838366 Apollo Scientific Ltd

2,6-Dibromo-4-fluorophenol

  • MDL number MFCD00002317
  • Cas Number : 344-20-7
  • Molecular Weight : 269.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-56
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-dibromo-4-fluorophenol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/26-dibromo-4-fluorophenol
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart