APO455838367 Apollo Scientific Ltd

1,3-Dichloro-2,4-difluorobenzene

  • MDL number MFCD00142519
  • Cas Number : 36556-37-3
  • Molecular Weight : 182.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,3-dichloro-2,4-difluorobenzene
Inchi Code 1S/C6H2Cl2F2/c7-3-1-2-4(9)5(8)6(3)10/h1-2H
Inchi Key XPHNTJXRSWNSNF-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/13-dichloro-24-difluorobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/13-dichloro-24-difluorobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart