APO455838368 Apollo Scientific Ltd

2,6-Dichloro-4-fluoroaniline

  • MDL number MFCD00142845
  • Cas Number : 344-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54-58
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/26-dichloro-4-fluoroaniline-97

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/26-dichloro-4-fluoroaniline-97
pictograms GHS06;GHS08;GHS09
hstatements H301+H311;H319;H410
Availability details for
Quantity
  • Availability
  • Add to Cart