APO455838369 Apollo Scientific Ltd

4,5-Dichloro-2-fluorobenzoic acid

  • MDL number MFCD00672988
  • Cas Number : 289039-49-2
  • Molecular Weight : 209

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-150
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/45-dichloro-2-fluorobenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/45-dichloro-2-fluorobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart