APO455838371 Apollo Scientific Ltd

4',5'-Difluoro-2'-nitroacetanilide

  • MDL number MFCD00143199
  • Cas Number : 1662-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3098

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=3098
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart