APO455838372 Apollo Scientific Ltd

4,5-Dimethoxy-2-fluorobenzonitrile

  • MDL number MFCD00143236
  • Cas Number : 119396-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3149

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=3149
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart