APO455838377 Apollo Scientific Ltd

4-Fluoro-2-methoxybenzaldehyde

  • MDL number MFCD00143318
  • Cas Number : 450-83-9
  • Molecular Weight : 154.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57-60
Synonym 4-Fluoro-o-anisaldehyde
Purity 98
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-2-methoxybenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-2-methoxybenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart