APO455838378 Apollo Scientific Ltd

2-Fluoro-4-methoxybenzoic acid

  • MDL number MFCD00509373
  • Cas Number : 394-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 193
Synonym 4-Carboxy-3-fluoroanisole, 2-Fluoro-p-anisic acid
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-4-methoxybenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-4-methoxybenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart