APO455838379 Apollo Scientific Ltd

2-Fluoro-5-methoxybenzoic acid

  • MDL number MFCD00272566
  • Cas Number : 367-83-9
  • Molecular Weight : 170.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146-150
Synonym 3-Carboxy-4-fluoroanisole, 6-Fluoro-m-anisic acid
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-5-methoxybenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-5-methoxybenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart