APO455838384 Apollo Scientific Ltd

4-(Trifluoromethoxy)benzotrifluoride

  • MDL number MFCD00190125
  • Cas Number : 80258-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5172

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=5172
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart