APO455838385 Apollo Scientific Ltd

4-Chloro-3-fluorobenzotrifluoride

  • MDL number MFCD00060646
  • Cas Number : 32137-20-5
  • Molecular Weight : 198.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Chloro-2-fluoro-4-(trifluoromethyl)benzene
Purity 99
Density 1.49
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 135
Product Link https://store.apolloscientific.co.uk/product/4-chloro-3-fluorobenzotrifluoride

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-3-fluorobenzotrifluoride
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart