APO455838388 Apollo Scientific Ltd

3-Fluoro-4-methylphenol

  • MDL number MFCD00143164
  • Cas Number : 452-78-8
  • Molecular Weight : 126.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-fluoro-4-methylphenol
Inchi Code 1S/C7H7FO/c1-5-2-3-6(9)4-7(5)8/h2-4,9H,1H3
Inchi Key GJOOCAXPERKNMN-UHFFFAOYSA-N

Description

Synonym 2-Fluoro-4-hydroxytoluene
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-4-methylphenol

Safety Information

signalword Danger
pcodes P280;P301+P312;P301+P330+P331
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-4-methylphenol
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart