APO455838396 Apollo Scientific Ltd

4-Bromo-2,6-difluorobenzyl bromide

  • MDL number MFCD03094462
  • Cas Number : 162744-60-7
  • Molecular Weight : 285.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-2-(bromomethyl)-1,3-difluorobenzene
density 1.991
Inchi Code 1S/C7H4Br2F2/c8-3-5-6(10)1-4(9)2-7(5)11/h1-2H,3H2
Inchi Key VSKMLLGUWKLTQV-UHFFFAOYSA-N

Description

Synonym 5-Bromo-2-(bromomethyl)-1,3-difluorobenzene, alpha,4-Dibromo-2,6-difluorotoluene
Purity 98%
Density 1.991
Flash Point 243
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-26-difluorobenzyl-bromide

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-26-difluorobenzyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart