APO455838397 Apollo Scientific Ltd

5-(Trifluoromethyl)thieno[3,2-b]pyridine-6-carboxylic acid

  • MDL number MFCD00203137
  • Cas Number : 243977-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205-208
Purity 95
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5509

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=5509
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart