APO455838407 Apollo Scientific Ltd

4-Amino-2,3,5-trifluoropyridine

  • MDL number MFCD01862081
  • Cas Number : 105252-95-7
  • Molecular Weight : 148.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 103-105
Iupac Name 2,3,5-trifluoro-4-pyridinamine
Inchi Code 1S/C5H3F3N2/c6-2-1-10-5(8)3(7)4(2)9/h1H,(H2,9,10)
Inchi Key YGXNVYOBTXUTMF-UHFFFAOYSA-N

Description

Synonym 2,3,5-Trifluoropyridin-4-amine
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 103-105
Product Link https://store.apolloscientific.co.uk/product/4-amino-235-trifluoropyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-235-trifluoropyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart