APO455838409 Apollo Scientific Ltd

4-Amino-2,5-difluorophenol

  • MDL number MFCD03094492
  • Cas Number : 120103-19-7
  • Molecular Weight : 145.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-130
Synonym 2,5-Difluoro-4-hydroxyaniline
Flash Point >110
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-25-difluorophenol

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-25-difluorophenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart