APO455838414 Apollo Scientific Ltd

1-Bromo-2,3,6-trifluorobenzene

  • MDL number MFCD00236214
  • Cas Number : 176793-04-7
  • Molecular Weight : 210.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point 62
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-bromo-236-trifluorobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-bromo-236-trifluorobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart