APO455838416 Apollo Scientific Ltd

4-[2-Nitro-4-(trifluoromethyl)phenyl]homopiperazine, N1-BOC protected

  • MDL number MFCD00729028
  • Cas Number : 306934-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72-73
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2525

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=2525
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart