APO455838417 Apollo Scientific Ltd

2-Amino-5-(3,4,4-trifluorobut-3-en-1-ylthio)-1,3,4-thiadiazole

  • MDL number MFCD01443829
  • Cas Number : 145070-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-162
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6209

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=6209
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart