APO455838418 Apollo Scientific Ltd

3-Amino-1,1,1-trifluoropropan-2-ol

  • MDL number MFCD02082679
  • Cas Number : 431-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99
Synonym 2-Hydroxy-3,3,3-trifluoropropylamine, 3-Amino-2-hydroxy-1,1,1-trifluoropropane
Flash Point >110
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-amino-111-trifluoropropan-2-ol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-111-trifluoropropan-2-ol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart