APO455838420 Apollo Scientific Ltd

3-Cyano-2-fluorobenzeneboronic acid

  • MDL number MFCD07374984
  • Cas Number : 957121-05-0
  • Molecular Weight : 164.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Borono-2-fluorobenzonitrile
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-cyano-2-fluorobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-cyano-2-fluorobenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart